accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3ON

CAS RN 202821-16-7

IUPAC Name: 2-(2-tert-butylphenoxy)acetonitrile
Synonyms: 2-(2-tert-butylphenoxy)acetonitrile
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: VYTLSISOMQEFSO-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC=C1OCC#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results