accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20283-92-5

OHO(R)HOOHOOOHOH(E)

CAS RN 20283-92-5

IUPAC Name: (2R)-3-(3,4-dihydroxyphenyl)-2-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}propanoic acid
Synonyms: rosemary acid (r-(e))-isomer rosmarinic acid (r-(e))-rosmarinic acid (R-(E))-isomer rosmarinic acid (R-(E))-rosmarinic acid (R)-rosmarinic acid
Molecular Weight (g/mol): 360.32
Molecular Formula: C18H16O8
InChi Key: DOUMFZQKYFQNTF-WUTVXBCWSA-N
SMILES: OC(=O)[C@@H](CC1=CC=C(O)C(O)=C1)OC(=O)\C=C\C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results