accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2030-63-9

IUPAC Name: N,5-bis(4-chlorophenyl)-3-[(propan-2-yl)imino]-3,5-dihydrophenazin-2-amine
Synonyms: clofazimina
Molecular Weight (g/mol): 473.4
Molecular Formula: C27H22Cl2N4
InChi Key: WDQPAMHFFCXSNU-UHFFFAOYSA-N
SMILES: CC(C)N=C1C=C2N(C3=CC=C(Cl)C=C3)C3=CC=CC=C3N=C2C=C1NC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (7)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)