accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2031-67-6

IUPAC Name: triethoxy(methyl)silane
Synonyms: methyltriethoxylane
Molecular Weight (g/mol): 178.3
Molecular Formula: C7H18O3Si
InChi Key: CPUDPFPXCZDNGI-UHFFFAOYSA-N
SMILES: CCO[Si](C)(OCC)OCC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results