accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFFF

CAS RN 203245-17-4

IUPAC Name: 1,2,3-trifluoro-5-methoxybenzene
Synonyms: 1,2,3-trifluoro-5-methoxybenzene
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: WVGMXLCSROHOIY-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(F)C(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results