CAS RN 203312-38-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Spirotetramat Metabolite BYI08330-cis-enol, TRC
CAS: 203312-38-3 Molecular Formula: C18 H23 N O3 Molecular Weight (g/mol): 301.38 Synonym: cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]dec-3-en-2-one,Spirotetramat-enol IUPAC Name: 3-(2,5-dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]dec-3-en-2-one SMILES: CO[C@@H]1CC[C@@]2(CC1)NC(=O)C(=C2O)c3cc(C)ccc3C