accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20344-49-4

HOFeO

CAS RN 20344-49-4

IUPAC Name: ironoylol
Synonyms: iron hydroxide
Molecular Weight (g/mol): 88.85
Molecular Formula: FeHO2
InChi Key: AEIXRCIKZIZYPM-UHFFFAOYSA-M
SMILES: O[Fe]=O
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (1)
  • (9)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)