accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20349-89-7

IUPAC Name: methyl 3-(2-hydroxyphenyl)propanoate
Synonyms: methyl 3-(2-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: YHXYRISRGHSPNV-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results