accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2035-72-5

H3CCH2OOHOO

CAS RN 2035-72-5

IUPAC Name: 4-benzoyl-3-hydroxyphenyl 2-methylprop-2-enoate
Synonyms: 4-benzoyl-3-hydroxyphenyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 282.3
Molecular Formula: C17H14O4
InChi Key: IMNBHNRXUAJVQE-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results