accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3NOO

CAS RN 20357-25-9

IUPAC Name: 4,5-dimethoxy-2-nitrobenzaldehyde
Synonyms: 4,5-dimethoxy-2-nitrobenzaldehyde
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: YWSPWKXREVSQCA-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=C(C=C1OC)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)