accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOHH2O

CAS RN 203626-75-9

IUPAC Name: isoquinoline-3-carboxylic acid hydrate
Synonyms: isoquinoline-3-carboxylic acid hydrate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: IXFLTVWYYQIVRB-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=CC2=CC=CC=C2C=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results