accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SHCl

CAS RN 2037-31-2

IUPAC Name: 3-chlorobenzene-1-thiol
Synonyms: 3-chlorobenzenethiol
Molecular Weight (g/mol): 144.62
Molecular Formula: C6H5ClS
InChi Key: CQJDYPZUDYXHLM-UHFFFAOYSA-N
SMILES: SC1=CC=CC(Cl)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)