CAS RN 2039-76-1
IUPAC Name:
1-(phenanthren-3-yl)ethan-1-one
Synonyms:
1-(phenanthren-3-yl)ethanone
Molecular Weight (g/mol):
220.27
Molecular Formula:
C16H12O
InChi Key:
JKVNPRNAHRHQDD-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C2C=CC3=CC=CC=C3C2=C1
1
–
4
of
4
results