CAS RN 2039-90-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dimethylstyrene, TRC
CAS: 2039-90-9 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 Synonym: 2-Ethenyl-1,3-dimethylbenzene (ACI),Styrene, 2,6-dimethyl- (7CI, 8CI),1,3-Dimethyl-2-vinylbenzene,2,6-Dimethylstyrene,2-Vinyl-m-xylene SMILES: Cc1cccc(C)c1C=C