accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2041-14-7

NH2POHOHO

CAS RN 2041-14-7

IUPAC Name: (2-aminoethyl)phosphonic acid
Synonyms: ciliatine
Molecular Weight (g/mol): 125.06
Molecular Formula: C2H8NO3P
InChi Key: QQVDJLLNRSOCEL-UHFFFAOYSA-N
SMILES: NCCP(O)(O)=O
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results