accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OCuOH2

CAS RN 20427-59-2

IUPAC Name: copper(II) hydroxide
Synonyms: copper(II) hydroxide
Molecular Weight (g/mol): 99.58
Molecular Formula: CuH4O2
InChi Key: AEJIMXVJZFYIHN-UHFFFAOYSA-N
SMILES: O.O.[Cu]
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results