accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2043-06-3

IUPAC Name: bis({[1,1'-biphenyl]-4-yl})-1,3,4-oxadiazole
Synonyms: bis({[1,1'-biphenyl]-4-yl})-1,3,4-oxadiazole
Molecular Weight (g/mol): 374.44
Molecular Formula: C26H18N2O
InChi Key: IVVYVRLKDGGUEL-UHFFFAOYSA-N
SMILES: O1C(=NN=C1C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results