CAS RN 2043-47-2
IUPAC Name:
3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol
Synonyms:
3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol
Molecular Weight (g/mol):
264.09
Molecular Formula:
C6H5F9O
InChi Key:
JCMNMOBHVPONLD-UHFFFAOYSA-N
SMILES:
OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F