accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2044-56-6

IUPAC Name: lithium(1+) dodecyl sulfate
Synonyms: lithium(1+) dodecyl sulfate
Molecular Weight (g/mol): 272.33
Molecular Formula: C12H25LiO4S
InChi Key: YFVGRULMIQXYNE-UHFFFAOYSA-M
SMILES: [Li+].CCCCCCCCCCCCOS([O-])(=O)=O
Quantity 
  • (1)
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)