accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20469-63-0

IUPAC Name: 1-iodo-2,4-dimethoxybenzene
Synonyms: 1-iodo-2,4-dimethoxybenzene
Molecular Weight (g/mol): 264.06
Molecular Formula: C8H9IO2
InChi Key: ZDUYJCKEORTAQE-UHFFFAOYSA-N
SMILES: COC1=CC=C(I)C(OC)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results