accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NSHOO

CAS RN 20485-41-0

IUPAC Name: 4-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: 4-methyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 143.16
Molecular Formula: C5H5NO2S
InChi Key: ZGWGSEUMABQEMD-UHFFFAOYSA-N
SMILES: CC1=C(SC=N1)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results