accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CI

CAS RN 2050-77-3

IUPAC Name: 1-iododecane
Synonyms: 1-iododecane
Molecular Weight (g/mol): 268.18
Molecular Formula: C10H21I
InChi Key: SKIDNYUZJPMKFC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCI
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)