accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2052-01-9

IUPAC Name: 2-bromo-2-methylpropanoic acid
Synonyms: 2-bromo-2-methylpropanoic acid
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: XXSPGBOGLXKMDU-UHFFFAOYSA-N
SMILES: CC(C)(Br)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results