accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2052-07-5

IUPAC Name: 2-bromo-1,1'-biphenyl
Synonyms: O-bromobiphenyl
Molecular Weight (g/mol): 233.11
Molecular Formula: C12H9Br
InChi Key: KTADSLDAUJLZGL-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results