accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20526-97-0

IUPAC Name: (3Z)-3-[(4-chlorophenyl)methylidene]-1,3-dihydro-2-benzofuran-1-one
Synonyms: (3Z)-3-[(4-chlorophenyl)methylidene]-2-benzofuran-1-one
Molecular Weight (g/mol): 256.69
Molecular Formula: C15H9ClO2
InChi Key: OHRFHJYUEWVXBD-ZROIWOOFSA-N
SMILES: ClC1=CC=C(\C=C2/OC(=O)C3=CC=CC=C23)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results