CAS RN 20555-88-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N4-Hydroxycytosine (>90%), TRC
CAS: 20555-88-8 Molecular Formula: C4H5N3O2 Molecular Weight (g/mol): 127.1 Synonym: 6-(Hydroxyamino)-2(1H)-pyrimidinone IUPAC Name: 6-(hydroxyamino)-1H-pyrimidin-2-one SMILES: ONC1=CC=NC(=O)N1