accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NClNBr

CAS RN 205672-25-9

IUPAC Name: 5-bromo-2-chloropyrimidin-4-amine
Synonyms: 5-bromo-2-chloropyrimidin-4-amine
Molecular Weight (g/mol): 208.44
Molecular Formula: C4H3BrClN3
InChi Key: QOWALNIZDHZTSM-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=NC=C1Br
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results