accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20582-85-8

IUPAC Name: 4-(methylsulfanyl)butan-1-ol
Synonyms: 4-(methylsulfanyl)butan-1-ol
Molecular Weight (g/mol): 120.21
Molecular Formula: C5H12OS
InChi Key: JNTVUHZXIJFHAU-UHFFFAOYSA-N
SMILES: CSCCCCO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)