accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 206-44-0

CAS RN 206-44-0

IUPAC Name: fluoranthene
Synonyms: fluoranthene 1,2-benzacenaphthene 1,2-(1,8-naphthylene)benzene benzo(jk)fluorene idryl rcra waste number U120 1,2-(1,8-naphthylene)-benzene
Molecular Weight (g/mol): 202.26
Molecular Formula: C16H10
InChi Key: GVEPBJHOBDJJJI-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13
Molecular Weight (g/mol) 
  • (3)
  • (10)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (10)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)