accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2060-55-1

IUPAC Name: [2,2'-bithiophene]-5-carboxylic acid
Synonyms: [2,2'-bithiophene]-5-carboxylic acid
Molecular Weight (g/mol): 210.27
Molecular Formula: C9H6O2S2
InChi Key: PTNWHEHDBNNKEO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results