accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SN

CAS RN 2060-63-1

IUPAC Name: 1-benzothiophene-5-carbonitrile
Synonyms: 1-benzothiophene-5-carbonitrile
Molecular Weight (g/mol): 159.21
Molecular Formula: C9H5NS
InChi Key: IPJUOWGTGHKFKN-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2SC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results