accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20624-25-3

IUPAC Name: sodium (diethylcarbamothioyl)sulfanide trihydrate
Synonyms: sodium diethyldithiocarbamate trihydrate
Molecular Weight (g/mol): 225.3
Molecular Formula: C5H16NNaO3S2
InChi Key: WWGXHTXOZKVJDN-UHFFFAOYSA-M
SMILES: O.O.O.[Na+].CCN(CC)C([S-])=S
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)