accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 206362-80-3

BrFCl

CAS RN 206362-80-3

IUPAC Name: 4-(bromomethyl)-1-chloro-2-fluorobenzene
Synonyms: 4-(bromomethyl)-1-chloro-2-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: CNUYBEIHDWMLSD-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results