accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COO

CAS RN 20651-71-2

IUPAC Name: 4-butylbenzoate
Synonyms: 4-butylbenzoate
Molecular Weight (g/mol): 177.22
Molecular Formula: C11H13O2
InChi Key: JFKUBRAOUZEZSL-UHFFFAOYSA-M
SMILES: CCCCC1=CC=C(C=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)