accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNH2HCl

CAS RN 206752-36-5

IUPAC Name: [amino(phenyl)methylidene]azanium
Synonyms: benzamidine
Molecular Weight (g/mol): 121.16
Molecular Formula: C7H9N2
InChi Key: PXXJHWLDUBFPOL-UHFFFAOYSA-O
SMILES: NC(=[NH2+])C1=CC=CC=C1
Percent Purity 
  • (7)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)