accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 206761-68-4

H3CONCS

CAS RN 206761-68-4

IUPAC Name: 3-isothiocyanato-4-methoxy-1,1'-biphenyl
Synonyms: 3-isothiocyanato-4-methoxy-1,1'-biphenyl
Molecular Weight (g/mol): 241.31
Molecular Formula: C14H11NOS
InChi Key: TZRMLJYZKZTEFW-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1N=C=S)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results