accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CON

CAS RN 20686-66-2

IUPAC Name: 2-methylnaphtho[2,3-d][1,3]oxazole
Synonyms: 2-methylnaphtho[2,3-d][1,3]oxazole
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: WMIHEWJRNFUVNR-UHFFFAOYSA-N
SMILES: CC1=NC2=CC3=CC=CC=C3C=C2O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results