accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20689-03-6

IUPAC Name: 1-[6-(benzyloxy)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]ethane-1,2-diol
Synonyms: 1-[6-(benzyloxy)-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]ethane-1,2-diol
Molecular Weight (g/mol): 310.35
Molecular Formula: C16H22O6
InChi Key: NKCNUNUEQRYLNZ-UHFFFAOYNA-N
SMILES: CC1(C)OC2C(OCC3=CC=CC=C3)OC(C(O)CO)C2O1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)