accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20691-89-8

IUPAC Name: (1-methylpiperidin-4-yl)methanol
Synonyms: (1-methylpiperidin-4-yl)methanol
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: KJZLJGZZDNGGCA-UHFFFAOYSA-N
SMILES: CN1CCC(CO)CC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results