accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OMnONOOONOO

CAS RN 20694-39-7

IUPAC Name: manganese(2+) tetrahydrate dinitrate
Synonyms: manganese(2+) tetrahydrate dinitrate
Molecular Weight (g/mol): 251.01
Molecular Formula: H8MnN2O10
InChi Key: ALIMWUQMDCBYFM-UHFFFAOYSA-N
SMILES: O.O.O.O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)