accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 206986-87-0

IUPAC Name: sodium 4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoate hydrate
Synonyms: sodium 4-{4,7,11-trihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoate hydrate
Molecular Weight (g/mol): 448.58
Molecular Formula: C24H41NaO6
InChi Key: MUVVIYFKOVLQHL-UHFFFAOYNA-M
SMILES: O.[Na+].CC(CCC([O-])=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results