accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOOHOAgAgAg

CAS RN 206986-90-5

IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate trisilver
Synonyms: citric acid hydrate trisilver
Molecular Weight (g/mol): 533.74
Molecular Formula: C6H10Ag3O8
InChi Key: HGXIBFSXZZKKRV-UHFFFAOYSA-N
SMILES: O.[Ag].[Ag].[Ag].OC(=O)CC(O)(CC(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results