accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOHOHOHOOHHOHOOHOHOOCH3OHMe

CAS RN 20702-77-6

IUPAC Name: 1-(4-{[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-2,6-dihydroxyphenyl)-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
Synonyms: neohesperidin dichalcone neohesperidin dihydrochalcone
Molecular Weight (g/mol): 612.58
Molecular Formula: C28H36O15
InChi Key: ITVGXXMINPYUHD-CUVHLRMHSA-N
SMILES: COC1=CC=C(CCC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)C=C2O)C=C1O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results