accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHH2NHSH2OHCl

CAS RN 207121-46-8

IUPAC Name: 2-amino-3-sulfanylpropanoic acid hydrate hydrochloride
Synonyms: cysteine hydrate hydrochloride
Molecular Weight (g/mol): 175.63
Molecular Formula: C3H10ClNO3S
InChi Key: QIJRTFXNRTXDIP-UHFFFAOYNA-N
SMILES: O.Cl.NC(CS)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)