CAS RN 207122-15-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2,2',4,4',5,6'-Hexabromodiphenyl Ether, TRC
CAS: 207122-15-4 Molecular Formula: C12 H4 Br6 O Molecular Weight (g/mol): 643.58 Synonym: Benzene, 1,3,5-tribromo-2-(2,4,5-tribromophenoxy)-,1,3,5-Tribromo-2-(2,4,5-tribromophenoxy)benzene,2,2',4,4',5,6'-Hexabromodiphenyl ether,BDE 154,PBDE 154,PBDE No. 154 IUPAC Name: 1,2,4-tribromo-5-(2,4,6-tribromophenoxy)benzene SMILES: Brc1cc(Br)c(Oc2cc(Br)c(Br)cc2Br)c(Br)c1
2
PBDE 154 50 μg/mL in Nonane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality