accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOBr

CAS RN 20717-79-7

IUPAC Name: 1-bromonaphthalene-2-carboxylic acid
Synonyms: 1-bromonaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 251.08
Molecular Formula: C11H7BrO2
InChi Key: VUVIRKAVBZITDO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=CC=CC2=C1Br
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results