accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH2NFF

CAS RN 207346-42-7

IUPAC Name: methyl 2-amino-4,5-difluorobenzoate
Synonyms: methyl 2-amino-4,5-difluorobenzoate
Molecular Weight (g/mol): 187.15
Molecular Formula: C8H7F2NO2
InChi Key: VIAQNTRKUPBQKR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(F)C=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results