accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COO

CAS RN 207511-19-1

IUPAC Name: 6-methyl-4H-chromen-4-one hydrate
Synonyms: 6-methylchromen-4-one hydrate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: QWIQUUMSUBFRAX-UHFFFAOYSA-N
SMILES: O.CC1=CC=C2OC=CC(=O)C2=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results