accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 207611-87-8

IUPAC Name: 4,4,5,5-tetramethyl-2-[4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 406.14
Molecular Formula: C24H32B2O4
InChi Key: INNVTCAYPFZCAL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results