accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

RhClClCl

CAS RN 20765-98-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Quantity 
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (6)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (6)
  • (3)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (6)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)